siRNA Details
| Gene Symbol: ZNF788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027049.1 | 3 | 2340 | |||
| GenBank mRNA | AK128282.1 | - | 1 | 1397 | ||
| AK128700.1 | - | 3 | 2340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF788| Gene Symbol: ZNF788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027049.1 | 3 | 2340 | |||
| GenBank mRNA | AK128282.1 | - | 1 | 1397 | ||
| AK128700.1 | - | 3 | 2340 | |||