siRNA Details
| Gene Symbol: FAM86EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130740.1 | 4 | 2339 | |||
| NR_130741.1 | 4 | 1393 | ||||
| NR_130742.1 | 4 | 1343 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM86EP| Gene Symbol: FAM86EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130740.1 | 4 | 2339 | |||
| NR_130741.1 | 4 | 1393 | ||||
| NR_130742.1 | 4 | 1343 | ||||