siRNA Details
| Gene Symbol: C21orf62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024622.1 | 2 | 314 | |||
| NR_024623.1 | 2 | 302 | ||||
| GenBank mRNA | AK096601.1 | - | 2 | 302 | ||
| BC117397.1 | - | 2 | 196 | |||
| BC117399.1 | - | 2 | 208 | |||
| DA321673.1 | - | 2 | 264 | |||
| DA776905.1 | - | 2 | 302 | |||

C21orf62-AS1
Genomic Map