siRNA Details
| Gene Symbol: LOC100129083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256795.1 | NP_001243724.1 | 3 | 600 | ||
| GenBank mRNA | CN369197.1 | - | 3 | 601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129083| Gene Symbol: LOC100129083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256795.1 | NP_001243724.1 | 3 | 600 | ||
| GenBank mRNA | CN369197.1 | - | 3 | 601 | ||