siRNA Details
| Gene Symbol: LOC284412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029390.1 | 1 | 1665 | |||
| GenBank mRNA | AF289566.1 | - | 1 | 904 | ||
| BC045185.1 | - | 1 | 1672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284412| Gene Symbol: LOC284412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029390.1 | 1 | 1665 | |||
| GenBank mRNA | AF289566.1 | - | 1 | 904 | ||
| BC045185.1 | - | 1 | 1672 | |||