siRNA Details
| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 11 | 1143 | |||
| NR_110307.1 | 9 | 901 | ||||
| NR_110308.1 | 10 | 1073 | ||||
| GenBank mRNA | BC033546.1 | - | 10 | 1044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00238| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 11 | 1143 | |||
| NR_110307.1 | 9 | 901 | ||||
| NR_110308.1 | 10 | 1073 | ||||
| GenBank mRNA | BC033546.1 | - | 10 | 1044 | ||