siRNA Details
| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 6 | 3265 | |||
| NR_027429.1 | 5 | 3178 | ||||
| NR_027430.1 | 5 | 3054 | ||||
| NR_027431.2 | 5 | 3028 | ||||
| NR_027432.1 | 4 | 2941 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM254-AS1| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 6 | 3265 | |||
| NR_027429.1 | 5 | 3178 | ||||
| NR_027430.1 | 5 | 3054 | ||||
| NR_027431.2 | 5 | 3028 | ||||
| NR_027432.1 | 4 | 2941 | ||||