This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DTX2P1-UPK3BP1-PMS2P11 SIRNA Assays ›
Gene Symbol:
DTX2P1-UPK3BP1-PMS2P11 LOC105375355
Gene Name:
DTX2P1-UPK3BP1-PMS2P11 readthrough, transcribed pseudogene
uroplakin-3b
Gene Aliases:
PMS2L11
-
Chromosome Location:
Chr.7: 76980822 - 77023761 on Build GRCh38
Chromosome Location:
Chr.7: 76510102 - 76524676 on Build GRCh38
UniGene ID:
Hs.675888
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID n269708

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DTX2P1-UPK3BP1-PMS2P11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_023383.1 6, 7 1092
GenBank mRNA BM709868.1 - 407
DR007973.1 - 1, 2 256
U38980.1 - 6, 7 978
Gene Symbol: LOC105375355
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005250613.4 XP_005250670.2 3, 4 766
XM_017012885.1 XP_016868374.1 3, 4 766
XM_017012886.1 XP_016868375.1 3, 4 766
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000579700.1 - 6, 7 812 Chr. 7 - 76981102 - 77043775 [+] on Build GRCh38
GENCODE ENST00000584900.5 - 6, 7 965 Chr. 7 - 76980949 - 77023761 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0