siRNA Details
| Gene Symbol: LINC01622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027115.3 | 5 | 2261 | |||
| NR_027116.2 | 4 | 2154 | ||||
| GenBank mRNA | BC037855.2 | - | 4 | 2155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01622| Gene Symbol: LINC01622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027115.3 | 5 | 2261 | |||
| NR_027116.2 | 4 | 2154 | ||||
| GenBank mRNA | BC037855.2 | - | 4 | 2155 | ||