siRNA Details
| Gene Symbol: LOC101929148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110413.1 | 2 | 372 | |||
| GenBank mRNA | AL133618.1 | - | 2 | 372 | ||
| Gene Symbol: TTTY6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA884105.1 | - | 1 | 56 | ||

LOC101929148
Genomic Map