siRNA Details
| Gene Symbol: LINC01548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024102.1 | 2 | 148 | |||
| GenBank mRNA | BC105115.1 | - | 2 | 148 | ||
| BC105117.1 | - | 2 | 148 | |||
| BX116511.1 | - | 2 | 101 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA400173.1 | - | 2 | 101 | Chr. 21 - 33165535 - 33170189 [-] on Build GRCh38 | |

LINC01548
Genomic Map