siRNA Details
| Gene Symbol: HYMAI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002768.2 | 1 | 1660 | |||
| GenBank mRNA | AF241534.1 | - | 1 | 1660 | ||
| BC059359.1 | - | 1 | 1181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HYMAI| Gene Symbol: HYMAI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002768.2 | 1 | 1660 | |||
| GenBank mRNA | AF241534.1 | - | 1 | 1660 | ||
| BC059359.1 | - | 1 | 1181 | |||