siRNA Details
| Gene Symbol: RMST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024037.1 | 8 | 952 | |||
| GenBank mRNA | AF429305.1 | - | 6 | 885 | ||
| AF429306.1 | - | 3 | 443 | |||
| AK056164.1 | - | 8 | 956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RMST| Gene Symbol: RMST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024037.1 | 8 | 952 | |||
| GenBank mRNA | AF429305.1 | - | 6 | 885 | ||
| AF429306.1 | - | 3 | 443 | |||
| AK056164.1 | - | 8 | 956 | |||