siRNA Details
| Gene Symbol: CNOT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005273481.2 | XP_005273538.1 | 8 | 5552 | ||
| GenBank mRNA | AK090522.1 | - | 1 | 1640 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK091357.1 | - | 1 | 816 | Chr. 8 - 17224967 - 17227277 [-] on Build GRCh38 | |

CNOT7
Genomic Map