siRNA Details
| Gene Symbol: FRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005266880.4 | XP_005266937.1 | 9 | 3344 | ||
| XM_005266881.2 | XP_005266938.1 | 9 | 3261 | |||
| XM_005266882.4 | XP_005266939.1 | 10 | 3573 | |||
| XM_011535653.2 | XP_011533955.1 | 10 | 3576 | |||
| XM_011535654.2 | XP_011533956.1 | 9 | 3358 | |||
| XM_011535655.2 | XP_011533957.1 | 9 | 3355 | |||
| XM_011535656.2 | XP_011533958.1 | 8 | 2729 | |||
| XM_017010645.1 | XP_016866134.1 | 9 | 3462 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK091699.1 | - | 2 | 1514 | Chr. 3 - 195693928 - 195696194 [+] on Build GRCh38 | |

FRK
Genomic Map