siRNA Details
| Gene Symbol: C1orf109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005270975.4 | XP_005271032.1 | 1 | 658 | ||
| XM_005270981.4 | XP_005271038.1 | 1 | 658 | |||
| XM_011541646.2 | XP_011539948.1 | 1 | 658 | |||
| XM_017001546.1 | XP_016857035.1 | 1 | 657 | |||
| GenBank mRNA | AK056029.1 | - | 1 | 1549 | ||

C1orf109
Genomic Map