siRNA Details
| Gene Symbol: LOC101928131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121677.1 | 4 | 888 | |||
| GenBank mRNA | BC039474.1 | - | 4 | 888 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928131| Gene Symbol: LOC101928131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121677.1 | 4 | 888 | |||
| GenBank mRNA | BC039474.1 | - | 4 | 888 | ||