siRNA Details
| Gene Symbol: LINC01355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110616.1 | 2 | 2782 | |||
| GenBank mRNA | AL832122.1 | - | 1 | 4715 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF677896.1 | - | 1 | 14 | Chr. 1 - 23281320 - 23282051 [-] on Build GRCh38 | |

LINC01355
Genomic Map