siRNA Details
| Gene Symbol: CTD-2194D22.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109912.1 | 4 | 1035 | |||
| GenBank mRNA | BC039383.1 | - | 4 | 1034 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000514569.1 | - | 4 | 1034 | Chr. 5 - 1887332 - 1900493 [+] on Build GRCh38 | |

CTD-2194D22.4
Genomic Map