siRNA Details
| Gene Symbol: GRIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648060.1 | - | 1 | 874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRIN2B| Gene Symbol: GRIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648060.1 | - | 1 | 874 | ||