siRNA Details
| Gene Symbol: LOC101928126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134561.1 | 2 | 833 | |||
| GenBank mRNA | AK124146.1 | - | 1 | 2157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928126| Gene Symbol: LOC101928126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134561.1 | 2 | 833 | |||
| GenBank mRNA | AK124146.1 | - | 1 | 2157 | ||