siRNA Details
| Gene Symbol: DLGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011541879.2 | XP_011540181.1 | 13 | 8603 | ||
| XM_011541880.2 | XP_011540182.1 | 8 | 6427 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK091069.1 | - | 1 | 1233 | Chr. 1 - 34861785 - 34863806 [-] on Build GRCh38 | |

DLGAP3
Genomic Map