siRNA Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142339.2 | NP_001135811.1 | 13 | 5449 | ||
| NM_001261443.1 | NP_001248372.1 | 13 | 5251 | |||
| NM_001261444.1 | NP_001248373.1 | 8 | 4969 | |||
| NM_182978.3 | NP_892023.1 | 12 | 5892 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR593590.1 | - | 1 | 1781 | Chr. 18 - 11883321 - 11885649 [+] on Build GRCh38 | |

GNAL
Genomic Map