siRNA Details
| Gene Symbol: ZNF862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054924.1 | - | 1 | 916 | ||
| BC045757.1 | - | 1 | 911 | |||
| BC092506.1 | - | 1 | 941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF862| Gene Symbol: ZNF862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054924.1 | - | 1 | 916 | ||
| BC045757.1 | - | 1 | 911 | |||
| BC092506.1 | - | 1 | 941 | |||