siRNA Details
| Gene Symbol: LOC101929384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120475.1 | 2 | 216 | |||
| GenBank mRNA | BC030099.2 | - | 2 | 216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929384| Gene Symbol: LOC101929384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120475.1 | 2 | 216 | |||
| GenBank mRNA | BC030099.2 | - | 2 | 216 | ||