siRNA Details
| Gene Symbol: LOC101929384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120475.1 | 3 | 561 | |||
| GenBank mRNA | BC030099.2 | - | 3 | 563 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG708751.1 | - | 3 | 564 | Chr. 12 - 203640 - 205007 [+] on Build GRCh38 | |

LOC101929384
Genomic Map