siRNA Details
| Gene Symbol: MCF2L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034002.1 | 1 | 1068 | |||
| GenBank mRNA | BC064982.1 | - | 1189 | |||
| BC080517.1 | - | 1189 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCF2L-AS1| Gene Symbol: MCF2L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034002.1 | 1 | 1068 | |||
| GenBank mRNA | BC064982.1 | - | 1189 | |||
| BC080517.1 | - | 1189 | ||||