siRNA Details
| Gene Symbol: ZMYM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190964.2 | NP_001177893.1 | 26 | 6602 | ||
| NM_001190965.2 | NP_001177894.1 | 25 | 6617 | |||
| NM_003453.4 | NP_003444.1 | 26 | 6710 | |||
| NM_197968.3 | NP_932072.1 | 25 | 6663 | |||
| XM_005266520.3 | XP_005266577.1 | 25 | 6622 | |||
| XM_011535216.2 | XP_011533518.1 | 26 | 6292 | |||
| XM_011535223.2 | XP_011533525.1 | 19 | 9007 | |||
| XM_011535224.2 | XP_011533526.1 | 19 | 4908 | |||
| XM_017020728.1 | XP_016876217.1 | 25 | 6553 | |||
| XM_017020729.1 | XP_016876218.1 | 25 | 6528 | |||
| XM_017020731.1 | XP_016876220.1 | 26 | 6345 | |||
| XM_017020732.1 | XP_016876221.1 | 26 | 6367 | |||
| XM_017020733.1 | XP_016876222.1 | 27 | 6394 | |||
| XM_017020734.1 | XP_016876223.1 | 27 | 6380 | |||
| XM_017020735.1 | XP_016876224.1 | 26 | 6344 | |||
| XM_017020736.1 | XP_016876225.1 | 26 | 6516 | |||
| XM_017020737.1 | XP_016876226.1 | 26 | 6291 | |||

ZMYM2
Genomic Map