siRNA Details
| Gene Symbol: LOC102723919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125443.1 | - | 2 | 2740 | ||
| Gene Symbol: LRIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005271369.2 | XP_005271426.1 | 17 | 11232 | ||
| XM_017002952.1 | XP_016858441.1 | 18 | 11204 | |||

LOC102723919
Genomic Map