siRNA Details
| Gene Symbol: ZNF891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277291.1 | NP_001264220.1 | 2 | 925 | ||
| XM_017018666.1 | XP_016874155.1 | 2 | 1093 | |||
| XM_017018667.1 | XP_016874156.1 | 2 | 763 | |||
| XM_017018668.1 | XP_016874157.1 | 2 | 846 | |||
| GenBank mRNA | BC017932.2 | - | 1 | 678 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG006010.1 | - | 1 | 411 | Chr. 12 - 133121217 - 133121743 [-] on Build GRCh38 | |

ZNF891
Genomic Map