siRNA Details
| Gene Symbol: LOC101928403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135540.1 | 1 | 761 | |||
| GenBank mRNA | BC017749.1 | - | 1 | 419 | ||
| BC046476.1 | - | 1 | 761 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928403| Gene Symbol: LOC101928403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135540.1 | 1 | 761 | |||
| GenBank mRNA | BC017749.1 | - | 1 | 419 | ||
| BC046476.1 | - | 1 | 761 | |||