siRNA Details
| Gene Symbol: LOC102606466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109872.1 | 2 | 570 | |||
| GenBank mRNA | BC047055.1 | - | 1 | 647 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102606466| Gene Symbol: LOC102606466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109872.1 | 2 | 570 | |||
| GenBank mRNA | BC047055.1 | - | 1 | 647 | ||