siRNA Details
| Gene Symbol: CCDC134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005261748.3 | XP_005261805.1 | 8 | 5911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC134| Gene Symbol: CCDC134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005261748.3 | XP_005261805.1 | 8 | 5911 | ||