siRNA Details
| Gene Symbol: LOC283588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055958.1 | - | 1 | 1590 | ||
| AK095276.1 | - | 1 | 2339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283588| Gene Symbol: LOC283588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055958.1 | - | 1 | 1590 | ||
| AK095276.1 | - | 1 | 2339 | |||