This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TTBK2 SIRNA Assays ›
Gene Symbol:
TTBK2
Gene Name:
tau tubulin kinase 2
Gene Aliases:
SCA11, TTBK
Chromosome Location:
Chr.15: 42738974 - 42920995 on Build GRCh38
UniGene ID:
Hs.646511
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n336676

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TTBK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005254171.4 XP_005254228.1 14 7436
XM_005254173.4 XP_005254230.1 14 7184
XM_006720402.3 XP_006720465.1 14 6911
XM_006720403.3 XP_006720466.1 15 7078
XM_017021950.1 XP_016877439.1 13 6927
GenBank mRNA AK074481.1 - 1 204
BX648356.1 - 1 1649

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

smoothened signaling pathway
regulation of cell shape
peptidyl-serine phosphorylation
cilium assembly
protein serine/threonine kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-q76w2:80/100.66.77.41:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0