This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC40A1 SIRNA Assays ›
Gene Symbol:
SLC40A1
Gene Name:
solute carrier family 40 member 1
Gene Aliases:
FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3
Chromosome Location:
Chr.2: 189560590 - 189580811 on Build GRCh38
UniGene ID:
Hs.643005
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n343487

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC40A1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017003938.1 XP_016859427.1 3 573
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK023574.1 - 1 1163 Chr. 2 - 189577633 - 189579519 [-] on Build GRCh38

Back To Top

More Information


Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

lymphocyte homeostasis
endothelium development
cellular iron ion homeostasis
regulation of transcription from RNA polymerase II promoter in response to iron
iron ion transmembrane transport
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
spleen trabecula formation
multicellular organismal iron ion homeostasis
ferrous iron export
receptor activity
iron ion transmembrane transporter activity
protein binding
ferrous iron transmembrane transporter activity
peptide hormone binding
iron channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0