siRNA Details
| Gene Symbol: LOC100287042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036520.1 | 1 | 2430 | |||
| GenBank mRNA | BC021162.2 | - | 1 | 1109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100287042| Gene Symbol: LOC100287042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036520.1 | 1 | 2430 | |||
| GenBank mRNA | BC021162.2 | - | 1 | 1109 | ||