This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC100506459 SIRNA Assays ›
Gene Symbol:
LOC100506459 CD58
Gene Name:
uncharacterized LOC100506459
CD58 molecule
Gene Aliases:
-
LFA-3, LFA3, ag3
Chromosome Location:
-
Chromosome Location:
Chr.1: 116487140 - 116571093 on Build GRCh38
UniGene ID:
Hs.734297
Hs.34341
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID n348106

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100506459
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126833.1 - 2 552
AL832882.1 - 1 2644
Gene Symbol: CD58
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017002869.1 XP_016858358.1 6 11238

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
single organismal cell-cell adhesion
heterotypic cell-cell adhesion
leukocyte migration
cellular response to interferon-gamma
cellular response to tumor necrosis factor
positive regulation of interleukin-8 secretion
receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4sh5g:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline