siRNA Details
| Gene Symbol: LOC101928053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120540.1 | 2 | 84 | |||
| GenBank mRNA | AK056207.1 | - | 2 | 84 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA701283.1 | - | 2 | 84 | Chr. 11 - 10809297 - 10821123 [+] on Build GRCh38 | |

LOC101928053
Genomic Map