siRNA Details
| Gene Symbol: HIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243198.2 | NP_001230127.1 | 29 | 7721 | ||
| NM_005338.6 | NP_005329.3 | 31 | 7874 | |||
| XM_005250304.2 | XP_005250361.1 | 31 | 7875 | |||
| XM_005250305.2 | XP_005250362.1 | 31 | 7880 | |||
| XM_011516116.2 | XP_011514418.1 | 32 | 7461 | |||
| XM_017012099.1 | XP_016867588.1 | 31 | 7795 | |||
| GenBank mRNA | BC082970.1 | - | 1 | 950 | ||

HIP1
Genomic Map