siRNA Details
| Gene Symbol: LOC101927770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110051.1 | 1 | 158 | |||
| GenBank mRNA | AK024093.1 | - | 1 | 158 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927770| Gene Symbol: LOC101927770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110051.1 | 1 | 158 | |||
| GenBank mRNA | AK024093.1 | - | 1 | 158 | ||