This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC101927770 SIRNA Assays ›
Gene Symbol:
LOC101927770
Gene Name:
uncharacterized LOC101927770
Gene Aliases:
-
Chromosome Location:
Chr.20: 53552770 - 53575308 on Build GRCh38
UniGene ID:
-
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n353814

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC101927770
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110051.1 6 1865
GenBank mRNA AK024093.1 - 6 1865

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-q76w2:80/100.66.77.41:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0