siRNA Details
| Gene Symbol: BEND5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096573.1 | - | 1 | 2145 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX116910.1 | - | 1 | 454 | Chr. 1 - 48728837 - 48729333 [-] on Build GRCh38 | |

BEND5
Genomic Map