siRNA Details
| Gene Symbol: CBR3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038894.1 | 3 | 521 | |||
| GenBank mRNA | AB004851.1 | - | 3 | 513 | ||
| BC038671.1 | - | 3 | 4442 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM730056.1 | - | 3 | 510 | Chr. 21 - 36141602 - 36156293 [-] on Build GRCh38 | |

CBR3-AS1
Genomic Map