siRNA Details
| Gene Symbol: LINC00693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038840.1 | 5 | 783 | |||
| GenBank mRNA | AA434563.1 | - | 3 | 319 | ||
| BX333929.2 | - | 3 | 463 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR624823.1 | - | 1 | 12 | Chr. 3 - 28757455 - 28758810 [+] on Build GRCh38 | |

LINC00693
Genomic Map