siRNA Details
| Gene Symbol: FLRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005267490.3 | XP_005267547.1 | 2 | 8023 | ||
| XM_017021129.1 | XP_016876618.1 | 2 | 8205 | |||
| XM_017021130.1 | XP_016876619.1 | 2 | 17479 | |||
| XM_017021131.1 | XP_016876620.1 | 3 | 16633 | |||
| XM_017021132.1 | XP_016876621.1 | 3 | 8112 | |||
| XM_017021133.1 | XP_016876622.1 | 3 | 8130 | |||
| XM_017021134.1 | XP_016876623.1 | 3 | 8057 | |||

FLRT2
Genomic Map