siRNA Details
| Gene Symbol: FP2234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF447871.1 | - | 1 | 1559 | ||
| AK091154.1 | - | 1 | 3559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FP2234| Gene Symbol: FP2234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF447871.1 | - | 1 | 1559 | ||
| AK091154.1 | - | 1 | 3559 | |||