This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CACNB2 SIRNA Assays ›
Gene Symbol:
CACNB2
Gene Name:
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases:
CACNLB2, CAVB2, MYSB
Chromosome Location:
Chr.10: 18140560 - 18545881 on Build GRCh38
UniGene ID:
Hs.59093
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n367370

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CACNB2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK128769.1 - 1 575

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline