siRNA Details
| Gene Symbol: LINC00384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648491.1 | - | 1 | 1108 | ||
| Gene Symbol: UTP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055279.1 | - | 3 | 930 | ||

LINC00384
Genomic Map