siRNA Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268492.2 | XP_005268549.1 | 3 | 13083 | ||
| XM_005268493.2 | XP_005268550.1 | 3 | 12479 | |||
| XM_006714789.2 | XP_006714852.1 | 3 | 13143 | |||
| XM_011537671.2 | XP_011535973.1 | 5 | 12978 | |||
| GenBank mRNA | AK091723.1 | - | 1 | 372 | ||
| BC036491.1 | - | 1 | 2189 | |||

KCTD16
Genomic Map